Formoterol Fumarate hydrate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

F0881-10MG F0881-50MG
TCIAF0881-10MGEA 608 DKK
TCIAF0881-10MG TCIAF0881-50MG
Formoterol Fumarate hydrate
Formoterol Fumarate hydrate
Formel: C₁₉H₂₄N₂O₄·½C₄H₄O₄·xH₂O
Opbevaringstemperatur: Fryser
MDL Number: MFCD03093863
CAS-nummer: 43229-80-7
Merck Index: 14,04245

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 % (total of 2peaks)
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 1.5 to 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR