3-Fluor-4-methoxyphenylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid

F0876-1G F0876-5G
TCIAF0876-1GEA 230 DKK
TCIAF0876-1G TCIAF0876-5G
3-Fluor-4-methoxyphenylboronsyre (contains varying amounts of Anhydride)
3-Fluor-4-methoxyphenylboronsyre
Formel: C₇H₈BFO₃
Smeltepunkt: 198°C
MDL Number: MFCD00807404
CAS-nummer: 149507-26-6

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR