Fmoc-D-Arg(Pbf)-OH ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-D-arginine, Fmoc-D-Arg(Pbf)-OH, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-D-arginine, Nalpha-Fmoc-Nomega-Pbf-D-arginine

F0875-1G F0875-5G
TCIAF0875-1GEA 140 DKK
TCIAF0875-1G TCIAF0875-5G
Fmoc-D-Arg(Pbf)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-D-Arg(Pbf)-OH
Formel: C₃₄H₄₀N₄O₇S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00237010
CAS-nummer: 187618-60-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +4.5 to +6.5 deg (C=1、DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR