1-Fluor-4-iod-3-nitrobenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Fluoro-1-iodo-2-nitrobenzene, 5-Fluoro-2-iodonitrobenzene, 2-Iodo-5-fluoronitrobenzene
F0828-5G
F0828-25G
TCIAF0828-5GEA
460
DKK
TCIAF0828-5G
TCIAF0828-25G
1-Fluor-4-iod-3-nitrobenzen ≥98.0% (by GC)
1-Fluor-4-iod-3-nitrobenzen
Formel:
C₆H₃FINO₂ Molekylvægt: 267 g/mol Kogepunkt: 128°C Smeltepunkt: 22.5 Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00153169 CAS-nummer: 364-77-2 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...