2-(4-Fluorophenyl)thiophene ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

F0724-1G F0724-5G
TCIAF0724-1GEA 255 DKK
TCIAF0724-1G TCIAF0724-5G
2-(4-Fluorophenyl)thiophene ≥95.0% (by GC)
2-(4-Fluorophenyl)thiophene
Formel: C₁₀H₇FS
MDL Number: MFCD06802535
CAS-nummer: 58861-48-6

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR