Fmoc-D-Glu-OH ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Nα-Fmoc-L-glutamic acid, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic acid, N-Fmoc-D-glutamic Acid
F0600-25G
F0600-5G
TCIAF0600-25GEA
862
DKK
TCIAF0600-25G
TCIAF0600-5G
Fmoc-D-Glu-OH ≥97.0% (by HPLC, titration analysis)
Fmoc-D-Glu-OH
Formel:
C₂₀H₁₉NO₆ |
MDL Number:
MFCD09037362 CAS-nummer: 104091-09-0 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | +18.0 to +22.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...