N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-Fmoc-D-methionine, Fmoc-D-Met-OH
F0594-1G
F0594-5G
TCIAF0594-1GEA
132
DKK
TCIAF0594-1G
TCIAF0594-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine
Formel:
C₂₀H₂₁NO₄S |
MDL Number:
MFCD00062958 CAS-nummer: 112883-40-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Specific rotation [a]20/D | +27.0 to +33.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...