FMOC-O-tert-butyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-α-Fmoc-o-t-butyl-L-Tyrosine, N-Fmoc-O-tert-butyl-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH
F0506-1G
F0506-25G
F0506-5G
TCIAF0506-1GEA
132
DKK
TCIAF0506-1G
TCIAF0506-25G
TCIAF0506-5G
FMOC-O-tert-butyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
FMOC-O-tert-butyl-L-tyrosine
Formel:
C₂₈H₂₉NO₅ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00037129 CAS-nummer: 71989-38-3 EINECS: 276-262-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Specific rotation [a]20/D | -28.0 to -32.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...