Fumaraldehydebis(dimethyl acetal) ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Fumaraldehyde bis(dimethylacetal), 1,1,4,4-Tetramethoxy-trans-2-butene, (E)-1,1,4,4-Tetramethoxybut-2-ene

F0329-1G F0329-5G
TCIAF0329-1GEA 353 DKK
TCIAF0329-1G TCIAF0329-5G
Fumaraldehydebis(dimethyl acetal) ≥90.0% (by GC)
Fumaraldehydebis(dimethyl acetal)
Formel: C₈H₁₆O₄
Molekylvægt: 176.21 g/mol
Flammepunkt: 80°C
MDL Number: MFCD00075507
CAS-nummer: 6068-62-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR