N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Fmoc-L-isoleucine, Fmoc-Ile-OH, FMOC-L-Isoleucine

F0294-25G F0294-5G
TCIAF0294-25GEA 723 DKK
TCIAF0294-25G TCIAF0294-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
Formel: C₂₁H₂₃NO₄
MDL Number: MFCD00037125
CAS-nummer: 71989-23-6
EINECS: 276-255-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 149.0 deg-C
Specific rotation [a]20/D -11.0 to -13.0 deg (C=1, DMF)
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR