3-(4-Fluorophenoxy)benzyl bromide ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3'-Bromomethyl-4-fluorodiphenyl Ether

F0243-1G F0243-5G
TCIAF0243-1GEA 1410 DKK
TCIAF0243-1G TCIAF0243-5G
3-(4-Fluorophenoxy)benzyl bromide ≥97.0% (by GC)
3-(4-Fluorophenoxy)benzyl bromide
Formel: C₁₃H₁₀BrFO
Molekylvægt: 281.12 g/mol
Kogepunkt: 160°C
MDL Number: MFCD00059915
CAS-nummer: 65295-58-1

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 97.0 %
Freezing point 30.0 to 37.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR