N-Formyl-L-leucine

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Formyl-Leu-OH, For-Leu-OH

F0131-1G F0131-5G
TCIAF0131-1GEA 255 DKK
TCIAF0131-1G TCIAF0131-5G
N-Formyl-L-leucine
N-Formyl-L-leucine
Formel: C₇H₁₃NO₃
Molekylvægt: 159.19 g/mol
MDL Number: MFCD00055861
CAS-nummer: 6113-61-7
EINECS: 228-080-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a754ed1c2fc4a756e0072ba0389a165c041951&vid=87a754ed1c2fc4a756e0072ba0389a165c041951&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 141.0 to 144.0 deg-C
Specific rotation [a]20/D -38.0 to -41.0 deg (C=1, H2O)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR