Fumaric acid di-(2-ethylhexyl) ester ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0117-25ML F0117-500ML
TCIAF0117-25MLEA 132 DKK
TCIAF0117-25ML TCIAF0117-500ML
Fumaric acid di-(2-ethylhexyl) ester ≥98.0% (by GC)
Fumaric acid di-(2-ethylhexyl) ester
Formel: C₂₀H₃₆O₄
Kogepunkt: 185°C
Flammepunkt: 190°C
MDL Number: MFCD00065144
CAS-nummer: 141-02-6
EINECS: 205-448-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85dc7b439dade9870ee08c2058f96b12c269a96c&vid=85dc7b439dade9870ee08c2058f96b12c269a96c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR