Dibutyl Fumarate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dibutyl fumarate, Fumaric Acid Dibutyl Ester

F0116-25G F0116-500G
TCIAF0116-25GEA 140 DKK
TCIAF0116-25G TCIAF0116-500G
Dibutyl Fumarate ≥98.0% (by GC)
Dibutyl Fumarate
Formel: C₁₂H₂₀O₄
Kogepunkt: 285°C
Smeltepunkt: –18°C
Flammepunkt: 142°C
MDL Number: MFCD00065141
CAS-nummer: 105-75-9
EINECS: 203-327-9

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR