Sodium hydrogen fumarate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Monosodium Fumarate, Fumaric Acid Monosodium Salt

F0071-25G F0071-500G
TCIAF0071-25GEA 132 DKK
TCIAF0071-25G TCIAF0071-500G
Sodium hydrogen fumarate ≥98.0% (by titrimetric analysis)
Sodium hydrogen fumarate
Formel: C₄H₄O₄
MDL Number: MFCD00039097
CAS-nummer: 5873-57-4
EINECS: 227-535-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR