2-Ethylhexyl palmitate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

E1435-250G E1435-25G
TCIAE1435-250GEA 854 DKK
TCIAE1435-250G TCIAE1435-25G
2-Ethylhexyl palmitate ≥98.0% (by GC)
2-Ethylhexyl palmitate
Formel: C₂₄H₄₈O₂
MDL Number: MFCD00072255
CAS-nummer: 29806-73-3
EINECS: 249-862-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.8550 to 0.8650


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR