1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Ethyl-4-(p-tolylethynyl)benzene, 1-Ethyl-4-[(p-tolyl)ethynyl]benzene, 1-Ethyl-4-[(4-methylphenyl)ethynyl]benzene, 1-(4-Ethylphenyl)-2-(p-tolyl)acetylene

E1408-25G E1408-5G
TCIAE1408-25GEA 2650 DKK
TCIAE1408-25G TCIAE1408-5G
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥98.0% (by GC)
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene
Formel: C₁₇H₁₆
MDL Number: MFCD04038793
CAS-nummer: 22692-80-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR