trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-Ethoxy-2,3-difluoro-4-(trans-4-pentylcyclohexyl)benzene
E1389-1G
E1389-5G
TCIAE1389-1GEA
206
DKK
TCIAE1389-1G
TCIAE1389-5G
trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)
trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene
Formel:
C₁₉H₂₈F₂O Molekylvægt: 310.43 g/mol |
MDL Number:
MFCD11053401 CAS-nummer: 124729-02-8 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...