Eltoprazine dihydrochloride ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(2,3-Dihydro-1,4-benzodioxin-5-yl)piperazine dihydrochloride

E1357-10MG E1357-50MG
TCIAE1357-10MGEA 862 DKK
TCIAE1357-10MG TCIAE1357-50MG
Eltoprazine dihydrochloride ≥98.0% (by HPLC)
Eltoprazine dihydrochloride
Formel: C₁₂H₁₆N₂O₂·₂HCl
CAS-nummer: 143485-51-2

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR