2-Ethylbenzoic acid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E1347-25G E1347-5G
TCIAE1347-25GEA 846 DKK
TCIAE1347-25G TCIAE1347-5G
2-Ethylbenzoic acid ≥98.0% (by GC)
2-Ethylbenzoic acid
Formel: C₉H₁₀O₂
Kogepunkt: 259°C
MDL Number: MFCD00045838
CAS-nummer: 612-19-1

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR