Ethylene Glycol Di(m-tolyl) ether ≥97.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 1,1'-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane
E1335-5G E1335-25G
TCIAE1335-5GEA 796 DKK
TCIAE1335-5G TCIAE1335-25G
Ethylene Glycol Di(m-tolyl) ether ≥97.0% (by GC)
Ethylene Glycol Di(m-tolyl) ether
Formel: C₁₆H₁₈O₂
Molekylvægt: 242.32 g/mol
MDL Number: MFCD00025952
CAS-nummer: 54914-85-1
EINECS: 402-730-9
UN nummer: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73d8b40396d8e50d69770ef0efc2834c50640a76&vid=73d8b40396d8e50d69770ef0efc2834c50640a76&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 96.0 to 100.0 deg-C