1-Fluor-3-ethynylbenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(3-Fluorophenyl)ethyne, 3-Fluorophenylacetylene, (3-Fluorophenyl)acetylene, 1-Ethynyl-3-fluorobenzene
E1332-5G
E1332-25G
TCIAE1332-5GEA
542
DKK
TCIAE1332-5G
TCIAE1332-25G
1-Fluor-3-ethynylbenzen ≥98.0% (by GC)
1-Fluor-3-ethynylbenzen
Formel:
C₈H₅F Kogepunkt: 40°C Flammepunkt: 32°C |
MDL Number:
MFCD03093798 CAS-nummer: 2561-17-3 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...