Ethyl 4-(trifluormethyl)benzoat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E1330-5G
E1330-25G
TCIAE1330-5GEA
247
DKK
TCIAE1330-5G
TCIAE1330-25G
Ethyl 4-(trifluormethyl)benzoat ≥98.0% (by GC)
Ethyl 4-(trifluormethyl)benzoat
Formel:
C₁₀H₉F₃O₂ Kogepunkt: 80°C |
MDL Number:
MFCD00013563 CAS-nummer: 583-02-8 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...