O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: HOTU, N-(6-Cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium hexafluorophosphate(V)

E1306-1G E1306-5G
TCIAE1306-1GEA 165 DKK
TCIAE1306-1G TCIAE1306-5G
O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate
Formel: C₁₀H₁₇F₆N₄O₃P
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01862832
CAS-nummer: 333717-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83e5575f3d438b362e91d942d895be8c9207cc75&vid=83e5575f3d438b362e91d942d895be8c9207cc75&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR