1,2-Ethylenediphosphonic acid ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

E1265-1G E1265-5G
TCIAE1265-1GEA 501 DKK
TCIAE1265-1G TCIAE1265-5G
1,2-Ethylenediphosphonic acid ≥97.0% (by titrimetric analysis)
1,2-Ethylenediphosphonic acid
Formel: C₂H₈O₆P₂
MDL Number: MFCD00013958
CAS-nummer: 6145-31-9
UN nummer: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbcf01e6e1772a4c59fb1de667e024fd14cc3838&vid=dbcf01e6e1772a4c59fb1de667e024fd14cc3838&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR