1-(Ethylamino)propan-2-ol ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

E1244-5ML E1244-25ML
TCIAE1244-5MLEA 723 DKK
TCIAE1244-5ML TCIAE1244-25ML
1-(Ethylamino)propan-2-ol ≥97.0% (by titrimetric analysis)
1-(Ethylamino)propan-2-ol
Formel: C₅H₁₃NO
Kogepunkt: 160°C
Smeltepunkt: 14°C
MDL Number: MFCD00082193
CAS-nummer: 40171-86-6
EINECS: 254-821-6
UN nummer: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee0a36d31659b2840ce0e0357fe1a6aa671b13df&vid=ee0a36d31659b2840ce0e0357fe1a6aa671b13df&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR