1-Ethyl-2-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
E1240-1G
E1240-5G
TCIAE1240-1GEA
403
DKK
TCIAE1240-1G
TCIAE1240-5G
1-Ethyl-2-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Ethyl-2-methylpyridinium bromide
Formel:
C₈H₁₂BrN |
MDL Number:
MFCD01208017 CAS-nummer: 32353-50-7 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...