Yoshimulactone Green (YLG) ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

E1238-10MG E1238-1MG
TCIAE1238-10MGEA 575 DKK
TCIAE1238-10MG TCIAE1238-1MG
Yoshimulactone Green (YLG) ≥98.0% (by HPLC)
Yoshimulactone Green (YLG)
Formel: C₂₇H₂₀O₇
CAS-nummer: 1807549-00-3

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 188.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR