1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

E1230-1G E1230-5G
TCIAE1230-1GEA 534 DKK
TCIAE1230-1G TCIAE1230-5G
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene
Formel: C₁₇H₂₄F₂O
Molekylvægt: 282.37 g/mol
MDL Number: MFCD11053400
CAS-nummer: 174350-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87879108f8daf05f7437b1e0588c9c26633223b3&vid=87879108f8daf05f7437b1e0588c9c26633223b3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 57.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR