(-)-Epicatechin ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2R,3R)-2-(3,4-Dihydroxyphenyl)chroman-3,5,7-triol, Epicatechol, (-)-cis-3,3',4',5,7-Pentahydroxyflavane

E1226-1G E1226-5G
TCIAE1226-1GEA 575 DKK
TCIAE1226-1G TCIAE1226-5G
(-)-Epicatechin ≥97.0% (by HPLC)
(-)-Epicatechin
Formel: C₁₅H₁₄O₆
Smeltepunkt: 240°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00075648
CAS-nummer: 490-46-0
EINECS: 207-710-1

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D -56.0 to -64.0 deg (C=0.1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR