Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: MCPB Ethyl ester, 4-(4-Chloro-2-methylphenoxy)butyric acid Ethyl ester

E1149-100MG E1149-1G
TCIAE1149-100MGEA 616 DKK
TCIAE1149-100MG TCIAE1149-1G
Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate ≥98.0% (by GC)
Ethyl 4-(4-Chloro-2-methylphenoxy)butyrate
Formel: C₁₃H₁₇ClO₃
Molekylvægt: 256.73 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00128924
CAS-nummer: 10443-70-6
UN nummer: 3082
ADR: 9 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR