7-(Ethylamino)-4,6-dimethylcoumarin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E1132-200MG E1132-1G
TCIAE1132-200MGEA 296 DKK
TCIAE1132-200MG TCIAE1132-1G
7-(Ethylamino)-4,6-dimethylcoumarin ≥98.0% (by GC)
7-(Ethylamino)-4,6-dimethylcoumarin
Formel: C₁₃H₁₅NO₂
MDL Number: MFCD00006860
CAS-nummer: 26078-25-1
EINECS: 247-446-4

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR