Ethyl 3-Methylpyrazole-5-carboxylate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ethyl 3-methyl-1H-pyrazole-5-carboxylate
E1100-1G
E1100-5G
TCIAE1100-1GEA
165
DKK
TCIAE1100-1G
TCIAE1100-5G
Ethyl 3-Methylpyrazole-5-carboxylate ≥98.0% (by GC)
Ethyl 3-Methylpyrazole-5-carboxylate
Formel:
C₇H₁₀N₂O₂ |
MDL Number:
MFCD00052514 CAS-nummer: 4027-57-0 EINECS: 223-702-0 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...