Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethyl 3-Carbamimidoylamino-p-toluate Nitrate, 3-Carbamimidoylamino-4-methylbenzoic Acid Ethyl Ester Nitrate, Ethyl 3-guanidino-4-methylbenzoate nitrate, 3-Carbamimidoylamino-p-toluic Acid Ethyl Ester Nitrate
E1079-1G
E1079-5G
TCIAE1079-1GEA
485
DKK
TCIAE1079-1G
TCIAE1079-5G
Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate ≥97.0% (by HPLC, titration analysis)
Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate
Formel:
C₁₁H₁₅N₃O₂ Molekylvægt: 284.27 g/mol |
MDL Number:
MFCD11100711 CAS-nummer: 641569-96-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 197.0 to 201.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...