Ethyl-1H-indazole-5-carboxylate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

E1067-1G E1067-5G
TCIAE1067-1GEA 591 DKK
TCIAE1067-1G TCIAE1067-5G
Ethyl-1H-indazole-5-carboxylate ≥98.0% (by GC)
Ethyl-1H-indazole-5-carboxylate
Formel: C₁₀H₁₀N₂O₂
MDL Number: MFCD03426220
CAS-nummer: 192944-51-7

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR