Ethyl-5-fluoro-1H-indole-2-carboxylate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethyl 5-fluoro-1H-indole-2-carboxylate, Ethyl 5-Fluoroindole-2-carboxylate, 5-Fluoroindole-2-carboxylic acid ethyl ester

E1035-1G E1035-5G
TCIAE1035-1GEA 600 DKK
TCIAE1035-1G TCIAE1035-5G
Ethyl-5-fluoro-1H-indole-2-carboxylate ≥98.0% (by GC)
Ethyl-5-fluoro-1H-indole-2-carboxylate
Formel: C₁₁H₁₀FNO₂
MDL Number: MFCD00152076
CAS-nummer: 348-36-7

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR