(4-Hydroxyphenyl)eddikesyreethylester ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Ethyl (4-hydroxyphenyl)acetate, Ethyl 2-(4-hydroxyphenyl)acetate
E1034-5G E1034-25G
TCIAE1034-5GEA 279 DKK
TCIAE1034-5G TCIAE1034-25G
(4-Hydroxyphenyl)eddikesyreethylester ≥98.0% (by GC)
(4-Hydroxyphenyl)eddikesyreethylester
Formel: C₁₀H₁₂O₃
Molekylvægt: 180.2 g/mol
Kogepunkt: 145°C
MDL Number: MFCD00016491
CAS-nummer: 17138-28-2
EINECS: 241-197-5
Bestil nu
Specifikationer Testresultater
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %