Ethyl-5H-octafluorovalerate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E1022-5G E1022-25G
TCIAE1022-5GEA 452 DKK
TCIAE1022-5G TCIAE1022-25G
Ethyl-5H-octafluorovalerate ≥98.0% (by GC)
Ethyl-5H-octafluorovalerate
Formel: C₇H₆F₈O₂
Kogepunkt: 180°C
MDL Number: MFCD00153153
CAS-nummer: 2795-50-8
UN nummer: 3272
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR