1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)
Leverandør: TCISamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 1-Boc-4-oxo-3-piperidinecarboxylate, Ethyl 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylate, 1-Boc-3-ethoxycarbonyl-4-piperidone, 1-tert-Butyl 3-ethyl 4-oxopiperidine-1,3-dicarboxylate, 1-Boc-4-oxo-3-piperidinecarboxylic acid ethyl ester, 1-tert-Butoxycarbonyl-3-ethoxycarbonyl-4-piperidone
Formel:
C₁₃H₂₁NO₅ Molekylvægt: 271.3100 g/mol |
MDL Number:
MFCD01862189 CAS-nummer: 98977-34-5 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...