Ethyl-5-nitrobenzofuran-2-carboxylate ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
5-Nitrobenzofuran-2-carboxylic Acid Ethyl Ester, Ethyl 5-nitrobenzofuran-2-carboxylate
E0950-5G
E0950-25G
TCIAE0950-5GEA
337
DKK
TCIAE0950-5G
TCIAE0950-25G
Ethyl-5-nitrobenzofuran-2-carboxylate ≥97.0% (by GC)
Ethyl-5-nitrobenzofuran-2-carboxylate
Formel:
C₁₁H₉NO₅ Molekylvægt: 235.20 g/mol |
MDL Number:
MFCD00275272 CAS-nummer: 69604-00-8 |
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...