4-Ethoxycinnamic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-(4-Ethoxyphenyl)acrylic Acid

E0856-25G E0856-5G
TCIAE0856-25GEA 3130 DKK
TCIAE0856-25G TCIAE0856-5G
4-Ethoxycinnamic acid ≥98.0% (by GC, titration analysis)
4-Ethoxycinnamic acid
Formel: C₁₁H₁₂O₃
Molekylvægt: 192.21 g/mol
MDL Number: MFCD11040581
CAS-nummer: 2373-79-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR