Ethyl-2-(p-Toluenesulphonyloxy)acetate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(p-Toluenesulphonyloxy)acetic Acid Ethyl Ester

E0851-1G E0851-5G
TCIAE0851-1GEA 1080 DKK
TCIAE0851-1G TCIAE0851-5G
Ethyl-2-(p-Toluenesulphonyloxy)acetate ≥98.0% (by GC)
Ethyl-2-(p-Toluenesulphonyloxy)acetate
Formel: C₁₁H₁₄O₅S
CAS-nummer: 39794-75-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR