Ethyl-trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester, trans-Ethyl 4-hydroxycyclohexanecarboxylate
E0838-5G
E0838-25G
TCIAE0838-5GEA
879
DKK
TCIAE0838-5G
TCIAE0838-25G
Ethyl-trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)
Ethyl-trans-4-hydroxycyclohexanecarboxylate
Formel:
C₉H₁₆O₃ Kogepunkt: 144°C |
MDL Number:
MFCD16295187 CAS-nummer: 3618-04-0 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...