Ethyl-7-chloro-2-oxoheptanoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

E0827-25G E0827-5G
TCIAE0827-25GEA 887 DKK
TCIAE0827-25G TCIAE0827-5G
Ethyl-7-chloro-2-oxoheptanoate ≥98.0% (by GC)
Ethyl-7-chloro-2-oxoheptanoate
Formel: C₉H₁₅ClO₃
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07698675
CAS-nummer: 78834-75-0
EINECS: 278-992-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR