Ethoxytriethylsilane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E0790-25ML E0790-5ML
TCIAE0790-25MLEA 887 DKK
TCIAE0790-25ML TCIAE0790-5ML
Ethoxytriethylsilane ≥98.0% (by GC)
Ethoxytriethylsilane
Formel: C₈H₂₀OSi
Kogepunkt: 155°C
MDL Number: MFCD00054718
CAS-nummer: 597-67-1
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR