7-Ethylcamptothecin ≥96.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
E0781-1G
E0781-100MG
TCIAE0781-1GEA
1860
DKK
TCIAE0781-1G
TCIAE0781-100MG
7-Ethylcamptothecin ≥96.0% (by HPLC)
7-Ethylcamptothecin
Formel:
C₂₂H₂₀N₂O₄ |
MDL Number:
MFCD06657922 CAS-nummer: 78287-27-1 UN nummer: 1544 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Specific rotation [a]20/D | +40.0 to +46.0 deg (C=0.4, CHCl3:MeOH=8:2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...