4-(Trifluoromethyl)phenylacetylene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Ethynylbenzotrifluoride, 4-Ethynyl-alpha,alpha,alpha-trifluorotoluene, 1-ethynyl-4-(trifluoromethyl)benzene
E0626-1G
E0626-5G
TCIAE0626-1GEA
575
DKK
TCIAE0626-1G
TCIAE0626-5G
4-(Trifluoromethyl)phenylacetylene ≥98.0% (by GC)
4-(Trifluoromethyl)phenylacetylene
Formel:
C₉H₅F₃ Molekylvægt: 170.134 g/mol Kogepunkt: 60°C Flammepunkt: 20°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01861903 CAS-nummer: 705-31-7 UN nummer: 1993 ADR: 3 II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...