4-Ethynylpyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4-Pyridylacetylene
E0561-100MG E0561-500MG
TCIAE0561-100MGEA 427 DKK
TCIAE0561-100MG TCIAE0561-500MG
4-Ethynylpyridine ≥98.0% (by GC, titration analysis)
4-Ethynylpyridine
Formel: C₇H₅N
Opbevaringstemperatur: Fryser
MDL Number: MFCD04038420
CAS-nummer: 2510-22-7
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 94.0 to 98.0 deg-C
Solubility in Acetone almost transparency