Ethyl isopropyl sulphone ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(Ethylsulphonyl)propane

E0475-25G E0475-5G
TCIAE0475-25GEA 1510 DKK
TCIAE0475-25G TCIAE0475-5G
Ethyl isopropyl sulphone ≥97.0% (by GC)
Ethyl isopropyl sulphone
Formel: C₅H₁₂O₂S
Kogepunkt: 91°C
MDL Number: MFCD00050418
CAS-nummer: 4853-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d881d77e81e3da7796f8b923f773b9dfb87258&vid=c8d881d77e81e3da7796f8b923f773b9dfb87258&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR