Ethyl methyl sulphone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethylmethylsulphone, Methylsulphonylethane

E0474-25G E0474-100G
TCIAE0474-25GEA 1050 DKK
TCIAE0474-25G TCIAE0474-100G
Ethyl methyl sulphone ≥98.0% (by GC)
Ethyl methyl sulphone
Formel: C₃H₈O₂S
Molekylvægt: 108.16 g/mol
MDL Number: MFCD00015543
CAS-nummer: 594-43-4

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR